Anti-FCER1 Armenian Hamster monoclonal antibody [clone: MAR-1]
Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40-5898-U100 40-5898-U500
40-5898-U100EA 866 DKK
40-5898-U100 40-5898-U500
Anti-FCER1 Armenian Hamster monoclonal antibody [clone: MAR-1]
Antistoffer
The MAR-1 antibody reacts with the Fc epsilon Receptor I alpha chain (FceRIa), a transmembrane protein member of the Ig superfamily. This chain, together with a beta chain and two gamma chains form a tetrameric complex that supports IgE-mediated signaling and subsequent release of chemical mediators of allergy and immediate hypersensitivity. FceR1a is upregulated in the presence of IgE on those cell types which express it, such as Mast cells and Basophils.
The MAR-1 antibody is widely used both in flow cytometry and for depletion of cells in vitro/in vivo. Please choose the appropriateMore Product Information