Ammoniakvand 10%, EMPROVE® EXPERT BiopharmaceuticalProduction, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ammoniak

4.80905.9200
4.80905.9200EA 0 DKK
4.80905.9200
Ammoniakvand 10%, EMPROVE® EXPERT BiopharmaceuticalProduction, SAFC®
Ammoniakvand
Formel: NH₃
Molekylvægt: 35.05 g/mol
Massefylde: 0.92 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00066650
CAS-nummer: 1336-21-6
EINECS: 215-647-6
UN nummer: 2672
ADR: 8 III

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR