Siliconcarbid 400 grinding compound

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

39800.LAEA 984 DKK
39800.LA
Siliconcarbid 400 grinding compound
Siliconcarbid

CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: Powder Density: 3.16 Appearance: Grey to black Odour: Odourless Melting Point: 2700° MDL No.: MFCD00049531


Grinding compound for polishing IR crystal windows

Formel: SiC
Molekylvægt: 40.1 g/mol
Smeltepunkt: 2700
Massefylde: 3.16
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00049531
CAS-nummer: 409-21-2
EINECS: 206-991-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR