96-brønds celledyrkningsplader, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3879
392-0291EA 1840 DKK
392-0291
96-brønds celledyrkningsplader, Corning®
Mikroplader Celledyrkningsplader
PS, klar, med låg.
  • Corning® CellBIND® surface increases wetability for more even and consistent cell attachment
  • Ultra-Low Attachment plates feature a covalently bound hydrogel layer that minimises cell attachment, protein absorption and cellular activation
  • Sterilised by gamma irradiation
  • Certified non-pyrogenic
Recommended working volume: 75 to 200 µl.