Borstandard, 1g/l B i vand (from B(OH₃)) Specpure® enkeltelement standard til ICP

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

39147.AD 39147.AE 39147.AP
39147.ADEA 699 DKK
39147.AD 39147.AE 39147.AP
Borstandard, 1g/l B i vand (from B(OH₃)) Specpure® enkeltelement standard til ICP
Standarder Kationiske standarder Boron Standards

Molecular Formula: H₃BO₃ in H₂O
Physical Form: Liquid
MDL No.: MFCD00134034


Certificeringer: Traceable to NIST.

Bestil nu

Specifikationer

Environmentally Preferable
MDL No. MFCD00134034

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR