Bacterial cell spreaders

Leverandør: Simport Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L300
391-3015EA 998 DKK
391-3015
Bacterial cell spreaders
Spredere Celle Spredere
PS, steril.

  • No flame sterilisation needed
  • Designed for easy spreading of cells onto the surface of an agar plate
  • Supplied sterile in individual packs

Handle length: 176 mm
Head spreader width: 36 mm
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR