Pierce™ Sealing Tape for 96-Well Microplates
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
15036
391-0649EA 1020 DKK
391-0649
Pierce™ Sealing Tape for 96-Well Microplates
Forsegling Mikrotiterpladetætninger
Microplate sealing tape consisting of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well or 384-well microplates during incubation steps.
  • Pressure-sensitive, just position the tape over plate and press to seal
  • Each sheet of tape is supplied on its own easy-to-peel backing
  • Durable and inert