VWR®, Versatile Cap, Smart barb- og taphane-teknologi
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-0644EA 829 DKK
216-0644 216-0645 216-0646 216-0647 216-0648 216-0649 216-0683 216-0684 216-0685 216-0686 216-0687 216-0688 216-0689 216-0690 216-0691 216-0692 216-0693 216-0694 216-0695 216-0696 216-0697 216-0698 391-0349 391-0350 391-0351 391-0352 391-0353 391-0354 391-0355
VWR®, Versatile Cap, Smart barb- og taphane-teknologi
Flasker Flasker, dunke
VWR® has expanded the VWR® carboy features and benefits by offering the Versatile Cap, Versatile Barb and state-of-the-art Spigot technology. These three components complement a wide array of applications supported by the VWR® Carboy. The leakproof cap insert with replaceable gasket enable users to quickly change the top connector of each carboy to match the application needs. Versatile Cap is stackable and is offered in both 83 mm (compatible with most industry carboys) and a 120 mm extra wide-mouth version that fits exclusively on VWR® 20 L and 40 L extra wide mouth carboys. To seeMore Product Information