CO₂ inkubator med styreskinner til WHEATON® rulleapparat, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WI057608-C-E
390-1588EA 274230 DKK
390-1588
CO₂ inkubator med styreskinner til WHEATON® rulleapparat, WHEATON®
Inkubatorer CO2-inkubatorer
WHEATON® CO₂ Incubators with an IR CO₂ sensor are ideal for everyday cell culture work and designed to be used with the WHEATON® Roller Apparatus.
  • Guide tracks in base for ease of roller apparatus entry
  • User configurable shelving - up to 30 shelves
  • Designed for temperature uniformity
  • Recovery time after 30 seconds door opening at 37 °C is less than 5 minutes
  • CO₂ recovery time after 30 seconds door opening at 5 Vol.% CO₂ is less than 5 minutes
  • Large viewing window in the door (with an option for dual windows)
  • Infrared sensor technology to maintain 0 to 20%