Formaldehyd 38% aqueous solution, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

3859.5000EA 0 DKK
3859.5000
Formaldehyd 38% aqueous solution, J.T.Baker®
Formaldehyd
Formel: CH₂O
Molekylvægt: 30.03 g/mol
Kogepunkt: -19.3
Smeltepunkt: -92
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003274
CAS-nummer: 50-00-0
EINECS: 200-001-8
UN nummer: 2209
ADR: 8 III
Merck Index: 13,04259

Bestil nu

Specifikationer Testresultater

Formaldehyde (HCHO)(after dilution) 36 - 40 g/l
pH 25°C (after dilution) 6.8 - 7.2
Total Phosphate max. 150 mM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR