Glassy carbon, plate, Thickness: 4 mm (0.16 in) Type 1

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

38026.FI 38026.GH 38026.FF
38026.FIEA 5480 DKK
38026.FI 38026.GH 38026.FF
Glassy carbon, plate, Thickness: 4 mm (0.16 in) Type 1
Glassy carbon
Formel: C
Molekylvægt: 12.01 g/mol
Smeltepunkt: 3652
Massefylde: 1.95 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133992
CAS-nummer: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR