1,4-Bis(methylamino)-9,10-anthraquinone 97% Dye, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,4-Bis(methylamino)anthraquinone, Disperse Blue 14, 1,4-Bis(methylamino)anthracene-9,10-dione

MQ100

367648-5G
367648-5GEA 768 DKK
367648-5G
1,4-Bis(methylamino)-9,10-anthraquinone 97% Dye, Sigma-Aldrich®
1,4-Bis(methylamino)-9,10-anthraquinone
Formel: C₁₆H₁₄N₂O₂
Molekylvægt: 266.29 g/mol
Smeltepunkt: 220...222 °C (lit.)
Opbevaringstemperatur: Room Temperature
MDL Number: MFCD00001198
CAS-nummer: 2475-44-7
EINECS: 219-602-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88753db8487e0274f4151b0c69dbb71410a8b78c&vid=88753db8487e0274f4151b0c69dbb71410a8b78c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance (Colour) Purple to Very Dark Purple
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength, C=0.01 G/L Ethanol 641 - 645 nm
Extinction Coefficient ≥14500
Wavelength, In Ethanol 593 - 597 nm
Purity (Titration by HClO4) 96.5 - 103.5 %
TLC Consistent with Past Lots
Solubility (Colour), c = 10 mg/ml; CH3CN Dark Blue to Very Dark Blue

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR