Ground & Waste Water Pollution Standard Solution 3, Specpure®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

36750.AEEA 2280 DKK
36750.AE
Ground & Waste Water Pollution Standard Solution 3, Specpure®
Standarder Miljø analyse standarder

For US EPA Clean Water Act.


  • 6 Analytes


UN No.: 3264
MDL No.: MFCD02091710

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR