Calcium phosphate tribasic (34 - 40% Ca)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Hydroxylapatite, tri-Calcium phosphate, HAp, Calcium Hydroxide Phosphate, Calcium hydroxyphosphate, Tribasic calcium phosphate

36731.36 36731.A3 36731.A9
36731.36EA 769 DKK
36731.36 36731.A3 36731.A9
Calcium phosphate tribasic (34 - 40% Ca)
Calcium phosphate tribasic

CAS No.: 12167-74-7 Molecular Formula: Ca10(OH)2(PO4)6 (approximate) Formula Weight: 1004.67 Physical Form: Powder Odour: Odourless MDL No.: MFCD00010904

Formel: Ca₅HO₁₃P₃
Molekylvægt: 502.306 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010904
CAS-nummer: 12167-74-7
EINECS: 235-330-6

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR