Salpetersyre solution_standardised 1.0 N

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Nitric acid solution

35624.AP 35624.K2
35624.APEA 507 DKK
35624.AP 35624.K2
Salpetersyre solution_standardised 1.0 N
Salpetersyre

CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Appearance: Colourless UN No.: 3264 MDL No.: MFCD00011349

Formel: HNO₃
Molekylvægt: 63.01 g/mol
Kogepunkt: 87
Smeltepunkt: -42
Massefylde: 1.035 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011349
CAS-nummer: 7697-37-2
EINECS: 231-714-2
UN nummer: 3264
ADR: 8 III
Merck Index: 13,06610

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR