Ammoniakvand solution_standardised 0.1 N

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ammoniak

35576.K2
35576.K2EA 733 DKK
35576.K2
Ammoniakvand solution_standardised 0.1 N
Ammoniakvand
Formel: NH₃
Molekylvægt: 35.05 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00066650
CAS-nummer: 1336-21-6
EINECS: 215-647-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR