CLP Protocol Test Kit for ICP Analysis, Specpure®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

35256.APEA 6360 DKK
35256.AP 35259.AP 35406.KS 35420.AE 35421.AE 35422.AE 41330.AE 41331.AE 35417.BG 35418.BG 35419.BG
CLP Protocol Test Kit for ICP Analysis, Specpure®
Standarder Kationiske standarder Multi-Element Standards

This LabPak™ comes with all the necessary standard concentrates needed for the day to day operations to complete the EPA established CLP Protocol requirements. Minimal mixing and dilution of the standard concentrates make it easy by saving you time and money in preparing all the necessary CLP Protocol Standards.


UN No.: UN3264
MDL No.: MFCD00146473


This LabPak™ contains the following CLP Protocol Test Kits for analysis by ICP:


Bestillingsoplysninger: Instrument Calibration (ICAL) Standards Kit (35417)
Initial Calibration Verification (ICV) Standards Kit (35418)
Continuing Calibration Verification (CCV) Standards Kit (35419)
Contract Required Detection Limits (CRDL) Standard (35420)
Interference Check Standard A (ICS-A) Solution (35256) - (2 each)
Interference Check Standard AB-1 (ICS-AB1) Solution (35421)
Interference Check Standard AB-2 (ICS-AB2) Solution (41330)
Water Spiking Standard A (WSS-A) Solution (35422)
Water Spiking Standard B (WSS-B) Solution (41331)
Initial Calibration Blank/Continuing Calibration Blank 4 (ICB/CCB) Standards Kit (35259) - (4 each)

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR