1-Imino-4-(1-(4-aminophenyl)-1-(4-amino-3-methylphenyl)methyliden)-2,5-cyclohexadien HCl for microscopical staining

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline

3525.0025EA 397 DKK
3525.0025
1-Imino-4-(1-(4-aminophenyl)-1-(4-amino-3-methylphenyl)methyliden)-2,5-cyclohexadien HCl for microscopical staining
1-Imino-4-(1-(4-aminophenyl)-1-(4-amino-3-methylphenyl)methyliden)-2,5-cyclohexadien HCl
Formel: C₂₀H₂₀ClN₃
Molekylvægt: 337.85 g/mol
Smeltepunkt: 268 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012569
CAS-nummer: 632-99-5
EINECS: 209-321-2
Merck Index: 12,05686

Bestil nu

Specifikationer Testresultater

Suitable for microscopy Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR