Kaliumchromat 1/30 M K2CrO4 (0.1N) volumetric solution, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

35157-1L
35157-1LEA 1100 DKK
35157-1L
Kaliumchromat 1/30 M K2CrO4 (0.1N) volumetric solution, Fluka™
Potassium chromate
Formel: K₂CrO₄
Molekylvægt: 194.19 g/mol
Kogepunkt: 1000
Smeltepunkt: 985
Massefylde: 1 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011368
CAS-nummer: 7789-00-6
Merck Index: 12,07784

Bestil nu

Specifikationer Testresultater

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR