Kaliumhydroxid 0.5 M (0.5 N) in ethanol volumetric solution, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Potash caustic, Caustic potash

35115-1L 35115-2.5L 35115-500ML 35115-6X1L 35115-4X2.5L
35115-1LEA 620 DKK
35115-1L 35115-2.5L 35115-500ML 35115-6X1L 35115-4X2.5L
Kaliumhydroxid 0.5 M (0.5 N) in ethanol volumetric solution, Fluka™
Potassium hydroxide
Formel: KOH
Molekylvægt: 56.11 g/mol
Kogepunkt: 78 °C (1.013 hPa)
Smeltepunkt: 360
Massefylde: 0.84 g/cm3 (20 °C)
Flammepunkt: 11 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003553
CAS-nummer: 1310-58-3
EINECS: 215-181-3
UN nummer: 2924
ADR: 3 II
Merck Index: 12,07806

Bestil nu

Specifikationer Testresultater

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR