Hanus solution (Iodine bromine) 0.1 M IBr Volumetric, Iodine_value, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Iodine solution acc. to Hanus

35097-1L 35097-500ML
35097-1LEA 1680 DKK
35097-1L 35097-500ML
Hanus solution (Iodine bromine) 0.1 M IBr Volumetric, Iodine_value, Fluka™
Hanus solution (Iodine bromine)
Formel: IBr
Massefylde: 1.06 g/cm³
Flammepunkt: 40 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011353
UN nummer: 2789
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Assay 0.0985 - 0.1014 mol/l

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR