Vand, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydrogen oxide

34877-2.5L 34877-4X2.5L 34877-1L
34877-2.5LEA 896 DKK
34877-2.5L 34877-4X2.5L 34877-1L
Vand, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Vand

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities


Beilstein Number: 2050024

Formel: H₂O
Molekylvægt: 18.02 g/mol
Kogepunkt: 100 °C
Smeltepunkt: 0 °C
Massefylde: 0.9971 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestil nu

Specifikationer Testresultater

Non-volatile matter Max. 3 ppm
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
HPLC Gradient at 254 nm Max. 1 mAU
HPLC Gradient at 210 nm Max. 5 mAU
Chloride (Cl) Max. 0.01 ppm
Fluoride (F) Max. 0.01 ppm
Nitrate (NO₃) Max. 0.1 ppm
Sulfate (SO₄) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR