Dimethylsulfoxid ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

34869-1L 34869-2.5L 34869-6X1L 34869-4X4L 34869-100ML 34869-2L 34869-4X2.5L 34869-6X100ML
34869-1LEA 1210 DKK
34869-1L 34869-2.5L 34869-6X1L 34869-4X4L 34869-100ML 34869-2L 34869-4X2.5L HONC34869-6X100ML
Dimethylsulfoxid ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Dimethylsulfoxid

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Formel: (CH₃)₂SO
Molekylvægt: 78.14 g/mol
Kogepunkt: 189 °C
Smeltepunkt: 18.5 °C
Massefylde: 1.104 g/cm³ (20 °C)
Flammepunkt: 28.5 %(V)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (GC) Min. 99.70 %
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.2 %
Absorbance at 350 nm Max. 0.01
Absorbance at 300 nm Max. 0.10
Absorbance 280 nm Max. 0.30
Absorbance at 270 nm Max. 0.70
Vapor Pressure 0.42 mmHg ( 20 °C)
Explosion Limit 42 %, 63 °F
Autoignition Temperature 573 °F
Vapor Density 2.7 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR