E/E Histology, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethyl alcohol

3405.9010EA 0 DKK
3405.9010
E/E Histology, J.T.Baker®
Ethanol
Formel: C₂H₅OH
Molekylvægt: 46.07 g/mol
Kogepunkt: 78.3
Smeltepunkt: –114 °C
Flammepunkt: 12
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
UN nummer: 1170
ADR: 3 II
Merck Index: 12,03806

Bestil nu

Specifikationer Testresultater

Assay (C2H5OH) min. 96 % (v/v)
Calcium (Ca) max. 5 ppm
Iron (Fe) max. 1 ppm
Residue after Evaporation max. 0.0015 %
Substances Darkened by H2SO4 passes test
Titrable Acid (meq/g) max. 0.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR