Calciumcarbonat ≥99.0% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Marble

33295.22 33295.36
33295.22EA 394 DKK
33295.22 33295.36
Calciumcarbonat ≥99.0% ACS
Calcium carbonate

Purity: 99.0% min CAS No.: 471-34-1 Molecular Formula: CaCO3 Formula Weight: 100.09 Physical Form: Powder Density: 2.930 Refractive Index: 1.6583 Appearance: White Odour: Odourless Melting Point: 800° dec. Merck Reference: 14,1657 MDL No.: MFCD00010906

Formel: CaCO₃
Molekylvægt: 100.09 g/mol
Smeltepunkt: 800 °C (dec.)
Massefylde: 2.930
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010906
CAS-nummer: 471-34-1
EINECS: 207-439-9
Merck Index: 14,1657

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR