Bromthymol blå solution_SC ethanol Indicator_acid_base, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

32822-100ML
32822-100MLEA 537 DKK
32822-100ML
Bromthymol blå solution_SC ethanol Indicator_acid_base, Fluka™
Bromthymol blå
Formel: C₂₇H₂₈Br₂O₅S
Molekylvægt: 624.39 g/mol
Flammepunkt: 38 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005872
CAS-nummer: 76-59-5
UN nummer: 1170
ADR: 3 III
Merck Index: 13,01430

Bestil nu

Specifikationer Testresultater

Test of sensitivity complying
Change range (pH 5,8-7,4) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR