1,2-Dichlorethan, extra dry 99.8%, AcroSeal®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethylendichlorid

326841000 326840010 326840025
326841000.EA 697 DKK
326841000. 326840010. 326840025.
1,2-Dichlorethan, extra dry 99.8%, AcroSeal®
1,2-Dichloroethane
Formel: ClH₂CCH₂Cl
Molekylvægt: 98.96 g/mol
Kogepunkt: 81…85 °C (760 mmHg)
Smeltepunkt: –35 °C
Massefylde: 1.25 g/cm³
Flammepunkt: 13 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000963
CAS-nummer: 107-06-2
EINECS: 203-458-1
UN nummer: 1184
ADR: 3 II
Merck Index: 13,03831
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48789e88e6041156e33712fd7c0198d7a4910266&vid=4238623c54017660b83d3baa856a69cc1aa8ee6d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.75 %
Residue after evaporation ≤0.002 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR