Saltsyre 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution

Leverandør: VWR Chemicals
Ratings:
Gennemsntilig vurdering: 5.0 (Bedømmelse)

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Hydrochloric acid solution, Muriatic acid

31954.290EA 234 DKK
31954.290 31954.368 31954.404
Saltsyre 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Saltsyre
Formel: HCl
Molekylvægt: 36.46 g/mol
Massefylde: 1.02 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
UN nummer: 1789
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR