Eosin Y 5 wt, % in H₂O in aqueous solution, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

M-Clarity™ quality level = MQ100

318906-500ML
318906-500MLEA 673 DKK
318906-500ML
Eosin Y 5 wt, % in H₂O in aqueous solution, Sigma-Aldrich®
Eosin Y

Eosin Y, also known as Eosin yellowish is a widely used xanthene dye. It is a counterstain that is used to distinguish between various connective tissue fibers and matrices, and the cytoplasm of different types of cells. Eosin in combination with hematoxylin is useful to demonstrate the general histological architecture of a tissue.


Eosin Y is suitable for use in hematoxylin and eosin (H and E) staining procedures for various tissue samples like lung nodule and kidney sections.

Formel: C₂₀H₆Br₄Na₂O₅
Massefylde: 1,015 g/ml (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005040
CAS-nummer: 17372-87-1
EINECS: 241-409-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27e9b95b7a1bcedbf957c22abdd8702fd9a87925&vid=5b4e65a5d3b4bca1f0255e74a007bfca5a09cd21&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance (Colour) Red to Very Dark Red
Appearance (Form) Liquid
Wavelength, In Water and 1% Na2CO3 514 - 520 nm
Extinction Coefficient ≥14000
Wavelength 252 - 258 nm
Extinction Coefficient ≥33000

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR