Ethylenglycolmonomethylether (2-Methoxyethanol) ≥99.3% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methylglykol

31733.AK 31733.K2
31733.AKEA 372 DKK
31733.AK 31733.K2
Ethylenglycolmonomethylether (2-Methoxyethanol) ≥99.3% ACS
Ethylenglycolmonomethylether (2-Methoxyethanol)

Purity: 99.3+% CAS No.: 109-86-4 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.965 Refractive Index: 1.4025 Appearance: Colourless Odour: Characteristic pH: 4-7 Melting Point: -85° Boiling Point: 124-125° Solubility: Miscible with water, alcohol, glycerol, ether, acetone, DMF Beilstein Reference: 1731074 Merck Reference: 14,6038 UN No.: 1188 MDL No.: MFCD00002867

Formel: C₃H₈O₂
Kogepunkt: 124…125 °C
Smeltepunkt: –85 °C
Massefylde: 0,965
Flammepunkt: 46 °C (115 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002867
CAS-nummer: 109-86-4
EINECS: 203-713-7
UN nummer: 1188
ADR: 3,III
Merck Index: 14,6038

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR