Natriumhydroxid 1 mol/l (1 N) aqueous solution, AVS TITRINORM Reag. USP, NF Volumetric

Leverandør: VWR Chemicals
Ratings:
Gennemsntilig vurdering: 5.0 (Bedømmelse)

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Caustic soda

31627.368EA 415 DKK
31627.368 31627.404 31627.290
Natriumhydroxid 1 mol/l (1 N) aqueous solution, AVS TITRINORM Reag. USP, NF Volumetric
Sodium hydroxide
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: 1390
Smeltepunkt: 323
Massefylde: 1.04 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8 II
Merck Index: 14,08627

Bestil nu

Specifikationer Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
Conforms to Reag. Ph.Eur. Passes test
Conforms to Reag. USP-NF Passes test
Conforms to BDH 19139 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR