Carbondisulfid 99,9 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., ACS IR, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

31627-1L
31627-1LEA 903 DKK
31627-1L
Carbondisulfid 99,9 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., ACS IR, Riedel-de Haën™
Carbondisulfid

Beilstein Number: 1098293
RTECS Number: FF6650000
UNSPSC Code: 12190000

Formel: CS₂
Molekylvægt: 76.14 g/mol
Kogepunkt: 46 °C (1.013 hPa)
Smeltepunkt: –76 °C
Massefylde: 1.264 g/cm³ (20 °C)
Flammepunkt: –30 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011321
CAS-nummer: 75-15-0
EINECS: 200-843-6
UN nummer: 1131
ADR: 3 I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Vapor Density 2.67 (vs air)
Explosion Limit 50 %
Assay (GC) Min. 99.9 %
Boiling range 46 - 47 °C
Density (D 20/4) 1.262 - 1.263
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
H₂S + foreign org. S-comp. Max. 0.0001 %
Sulfate (SO₄) Max. 0.00025 %
Sulfite (as SO₂) Max. 0.001 %
Benzene Max. 0.001 %
Iodine consuming matters complying
APHA Max. 10
Autoignition Temperature 212 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR