Kaliumhydroxid 45% aqueous solution, BAKER ANALYZED®, Electronic Grade, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Potash caustic, Caustic potash

3144-03EA 1290 DKK
3144-03
Kaliumhydroxid 45% aqueous solution, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Potassium hydroxide
Formel: KOH
Molekylvægt: 56.11 g/mol
Kogepunkt: 1320
Smeltepunkt: 360
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003553
CAS-nummer: 1310-58-3
EINECS: 215-181-3
UN nummer: 1814
ADR: 8 II
Merck Index: 12,07806

Bestil nu

Specifikationer Testresultater

For Microelectronic Use 45.0 - 46.5 %
Assay (KOH) 45.0 - 46.5 %
Color (APHA) ≤10
Appearance Passes Test
Density (g/mL) at 25°C 1.45 - 1.47
Identification Passes Test
Insoluble Matter ≤30 ppm
Potassium Carbonate (K₂CO₃) ≤0.1 %
Ammonium Hydroxide Precipitate ≤30 ppm
Chloride (Cl) ≤30 ppm
Nitrogen Compounds (as N) ≤2 ppm
Phosphate (PO₄) ≤1 ppm
ACS - Sulfate (SO₄) ≤5 ppm
Trace Impurities - Aluminum (Al) ≤5.000 ppm
Trace Impurities - Chromium (Cr) ≤5.0 ppm
Trace Impurities - Copper (Cu) ≤0.050 ppm
Heavy Metals (as Ag) ≤3 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤2.000 ppm
Trace Impurities - Mercury (Hg) ≤0.10 ppm
Trace Impurities - Nickel (Ni) ≤1.000 ppm
Trace Impurities - Sodium (Na) ≤300.000 ppm
Trace Impurities - Zinc (Zn) ≤5.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR