Kaliumdisulfit ≥96% (by iodometry), powder, Puriss. p.a. analytical reagent, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dipotassium disulphite, Potassium pyrosulphite, Potassium metabisulphite, di-Potassium disulphite

31268-1KG 31268-2.5KG 31268-500G 31268-6X1KG
31268-1KGEA 678 DKK
31268-1KG 31268-2.5KG 31268-500G 31268-6X1KG
Kaliumdisulfit ≥96% (by iodometry), powder, Puriss. p.a. analytical reagent, Fluka™
Potassium disulphite
Formel: K₂O₅S₂
Molekylvægt: 222.33 g/mol
Smeltepunkt: 190 Dec.
Massefylde: 2.3 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00167605
CAS-nummer: 16731-55-8
EINECS: 240-795-3
Merck Index: 13,07729

Bestil nu

Specifikationer Testresultater

Assay (Iodometric) 96 - 101.0 %
Arsenic (As) Max. 0.0001 %
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.001 %
Chloride (Cl) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR