Ethylacetat ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Acetic acid ethyl ester

31063-18L 31063-1L 31063-2.5L 31063-45L 31063-4X2.5L 31063-7L 31063-200L
31063-18LEA 9840 DKK
31063-18L 31063-1L 31063-2.5L 31063-45L 31063-4X2.5L 31063-7L HONC31063-200L
Ethylacetat ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Ethylacetat

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Formel: CH₃COOC₂H₅
Molekylvægt: 88.11 g/mol
Kogepunkt: 77 °C (1.013 hPa)
Smeltepunkt: –84 °C
Massefylde: 0.901 g/cm³
Flammepunkt: 11.5 %(V)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009171
CAS-nummer: 141-78-6
EINECS: 205-500-4
UN nummer: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Autoignition Temperature 801 °F
Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Suitability for residue analysis max. 5 ng/l
Vapor Density 3 (20 °C, vs air)
Vapor Pressure 73 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR