Kviksølv(II)nitrat 0.01 mol/l (0.02 N) aqueous solution

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Mercury dinitrate, Mercuric nitrate

310274.1000EA 249 DKK
310274.1000
Kviksølv(II)nitrat 0.01 mol/l (0.02 N) aqueous solution
Kviksølv(II)nitrat
Formel: HgN₂O₆
Molekylvægt: 324.60 g/mol
Smeltepunkt: -38.9
Massefylde: 4.3
MDL Number: MFCD00011038
CAS-nummer: 10045-94-0
EINECS: 233-152-3
UN nummer: 3287
ADR: 6.1 II
Merck Index: 12,05933

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR