Dragendorff's reagens for analysis of alkaloids

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

30989.236EA 884 DKK
30989.236
Dragendorff's reagens for analysis of alkaloids
Dragendorff's reagens

Made of potassium bismuth iodide

Kogepunkt: 77
Massefylde: 1.2 g/cm³ (20 °C)
Flammepunkt: –4 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00283106
CAS-nummer: 39775-75-2
UN nummer: 3264
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR