HEPES, anhydrous 99,5 BiopharmaceuticalProduction

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

30487.297EA 2600 DKK
30487.297 30487.366 30487.468
HEPES, anhydrous 99,5 BiopharmaceuticalProduction
HEPES

HEPES

Formel: C₈H₁₈N₂O₄S
Molekylvægt: 238.31 g/mol
Smeltepunkt: 210…215 °C
Massefylde: 1.448 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006158
CAS-nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay Min. 99.5 %
Appearance White crystalline powder
Cell culture toxicity Passes test
DNases Not detected
Identification by IR Conforms to structure
Proteases Not detected
RNases Not detected
Solubility (1 mol/l) Clear.colourless.complete
pKa (20°C) 7.45 - 7.65
Heavy metals (as Pb) Max. 5 ppm
Ignition residue Max. 0.3 %
Water Max. 0.5 %
Ca (Calcium) Max. 5 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 5 ppm
Mn (Manganese) Max. 5 ppm
Absorbance (260 nm) (1 mol/l) Max. 0.10
Absorbance (280 nm) (1 mol/l) Max. 0.10
Bioburden Max. 100 CFU/g
Endotoxin (0,2 %) Max. 0.1 EU/mg

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR