Kaliumiodat 99.7-100.4%, Puriss. p.a. analytical reagent, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

30314-100G 30314-1KG 30314-250G 30314-25KG
30314-100GEA 491 DKK
30314-100G 30314-1KG 30314-250G 30314-25KG
Kaliumiodat 99.7-100.4%, Puriss. p.a. analytical reagent, Fluka™
Potassium iodate
Formel: KlO₃
Molekylvægt: 214 g/mol
Smeltepunkt: 560 °C
Massefylde: 3.93 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011406
CAS-nummer: 7758-05-6
EINECS: 231-831-9
UN nummer: 1479
ADR: 5.1 II
Merck Index: 13,07726

Bestil nu

Specifikationer Testresultater

Assay 99.7 - 100.4 %
Water insoluble matter Max. 0.005 %
Loss on drying (130°C) Max. 0.05 %
pH (5 %, 25 °C) 5.0 - 8.0
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.001 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chlorate, Chloride, Bromide, Bromate (as Cl) Max. 0.01 %
Total N Max. 0.002 %
Iodide (I) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR