Polyurethane surface sampling bags with PolySponge™, Whirl-Pak®
Leverandør: NASCO FORT ATKINSON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B01590WA
300-0672EA 4210 DKK
300-0672
Polyurethane surface sampling bags with PolySponge™, Whirl-Pak®
Poser Prøveposer
Make environmental surface sampling faster and easier with Whirl-Pak polyurethane surface sampling bags with premoistened sponge. The durable high-density polyurethane sponge is ready to use and resists tearing and fraying.
  • Each sponge is packed in a sterile, sealed write-on bag
  • Write-on label makes identifying samples easy and convenient
  • Sponge is hydrated with 10 ml of HiCap™ neutralizing broth
  • Terminally gamma processed to guarantee sterility (+1 Sterilization)
  • Opening Tyvek® Pull Tabs are perpendicular to the opening of the bag: Keeps your hands away