G418 disulfat solution 50 mg/ml

Leverandør: Corning
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

30-234-CI 30-234-CR
MDTC30-234-CIEA 4240 DKK
MDTC30-234-CI 30-234-CR
G418 disulfat solution 50 mg/ml
G418 disulfat

G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 µg/ml.


This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene. G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.


Advarsel: For Research or Further Manufacturing Use Only. Not for use in Diagnostic Procedures.

Formel: C₂₀H₄₄N₄O₁₈S₂
Molekylvægt: 692.72 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00058314
CAS-nummer: 108321-42-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR