Kieselgel, Pore size: 60 Å, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


M-Clarity™ quality level = MQ400

288624-1KG 288624-250G 288624-5KG
288624-1KGEA 1600 DKK
288624-1KG 288624-250G 288624-5KG
Kieselgel, Pore size: 60 Å, Supelco®
Silica gel
Formel: SiO₂
Molekylvægt: 60.08 g/mol
Kogepunkt: 2230
Smeltepunkt: 1710
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011232
CAS-nummer: 63231-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d49d3c6ff6a726715819e3052dc7f4eaf107acc&vid=fab2e215fa9fc64905b054772d96c32c3944b69c&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR