Natriumdihydrogenphosphat dihydrat USP, Ph. Eur.

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

28014.291EA 839 DKK
28014.291 28014.360 28014.462
Natriumdihydrogenphosphat dihydrat USP, Ph. Eur.
Natriumdihydrogenphosphat dihydrat

Appearance: White or almost white, powder or colourless crystals.


Advarsel: Not for use as Active Pharmaceutical Ingredient (API)

Formel: NaH₂PO₄·2H₂O
Kogepunkt: < 650 °C (1013 hPa)
Smeltepunkt: 60 °C
Massefylde: 1,91 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149209
CAS-nummer: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Bestil nu

Specifikationer Testresultater

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) ≤10 ppm
Residual solvents Unlikely by manuf.process
As (Arsenic) ≤2 ppm
Cd (Cadmium) ≤1 ppm
Co (Cobalt) ≤1 ppm
Hg (Mercury) ≤3 ppm
Ni (Nickel) ≤1 ppm
Pb (Lead) ≤4 ppm
V (Vanadium) ≤5 ppm
Conforms to Ph.Eur./USP/BP Passes test
Ph.Eur./BP specifications: .
Assay (dried substance) [Ph.Eur./BP] 98.0 - 100.5 %
Identification A Passes test USP
Identification B Passes test USP
Identification C Passes test Ph.Eur./BP
Appearance of solution Passes test Ph.Eur./BP
pH (5 %) [Ph.Eur./BP] 4.2 - 4.5
Reducing substances Passes test Ph.Eur./BP
Cl (Chloride) [Ph.Eur./BP] ≤200 ppm
SO4 (Sulphate) [Ph.Eur./BP] ≤300 ppm
Fe (Iron) [Ph.Eur./BP] ≤10 ppm
Loss on drying (130°C) [Ph.Eur./BP] 21.5 - 24.0 %
USP specifications: .
Assay (calculated on anhydrous) [USP] 98.0 - 103.0 %
Insoluble substances [USP] ≤0.2 %
Cl (Chloride) [USP] ≤0.014 %
SO4 (Sulphate) [USP] ≤0.15 %
Al, Ca and related elements Passes test USP
As (Arsenic) [USP] ≤8 ppm
pH (25°C; 1 g/20 ml) [USP] 4.1 - 4.5
Water determination [USP] 18.0 - 26.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR