Natriummetabisulfit ≥98.0%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Natriumdisulfit, di-Natriumdisulfit, Natriumpentaoxodisulfat

27920.295EA 437 DKK
27920.295 27920.460
Natriummetabisulfit ≥98.0%, AnalaR NORMAPUR® analytical reagent
Natriummetabisulfit
Formel: Na₂S₂O₅
Molekylvægt: 190.11 g/mol
Smeltepunkt: 150
Massefylde: 1.48 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00167602
CAS-nummer: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay Min. 98.0 %
Heavy metals (as Pb) Max. 10 ppm
Cl (Chloride) Max. 100 ppm
As (Arsenic) Max. 0.5 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 5 ppm
Zn (Zinc) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR