Polyethylen glycol 400, GPR RECTAPUR®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: PEG 400

26602.290EA 284 DKK
26602.290 26602.460 26602.320
Polyethylen glycol 400, GPR RECTAPUR®
Polyethylen glycol 400
Formel: H(OCH₂CH₂)nOH
Kogepunkt: >250 °C (1013 hPa)
Smeltepunkt: 4…8 °C
Massefylde: 1,13 g/cm³ (20 °C)
Flammepunkt: 245 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081839
CAS-nummer: 25322-68-3
EINECS: 500-038-2

Bestil nu

Specifikationer Testresultater

Identification Passes test
Acid value (mg KOH/g) Max. 0.2
Density (20/4) 1.120 - 1.130
Hydroxyl value 200 - 300


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR