Natriumhydroxid 30% in aqueous solution, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Caustic soda

2617.91000EA 0 DKK
2617.91000
Natriumhydroxid 30% in aqueous solution, J.T.Baker®
Sodium hydroxide
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: 119…135 °C (1013 hPa)
Smeltepunkt: 9…10 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8 II
Merck Index: 14,08627

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR