Methylethylketon ≥99%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethylmethylketon, Butanon

25641.297EA 372 DKK
25641.297 25641.322 25641.366 25641.468 25641.550
Methylethylketon ≥99%, GPR RECTAPUR®
Methylethylketon
Formel: C₂H₅COCH₃
Molekylvægt: 72.11 g/mol
Kogepunkt: 79.6 °C (1013 hPa)
Smeltepunkt: –86 °C
Massefylde: 0.8054 g/cm³ (20 °C)
Flammepunkt: –4 °C (DIN 51755 Part 1)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011648
CAS-nummer: 78-93-3
EINECS: 201-159-0
UN nummer: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay Min. 99 %
IR Spectrum Passes test
Boiling point 79 - 80 °C
Acidity (at filling) Max. 0.0005 meq/g
Density (20/4) 0.804 - 0.807
Evaporation residue Max. 20 ppm
Water Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR