Methylsalicylat 99, GPR RECTAPUR®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Wintergreen Oil, Methyl 2-hydroxybenzoate

25616.291EA 1230 DKK
25616.291
Methylsalicylat 99, GPR RECTAPUR®
Methylsalicylat
Formel: 2-(HO)C₆H₄CO₂CH₃
Molekylvægt: 152.15 g/mol
Kogepunkt: 432
Smeltepunkt: 100…101 °C
Massefylde: 1.184 g/cm³ (20 °C)
Flammepunkt: 99
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002214
CAS-nummer: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay Min. 99 %
Density (20/4) 1.180 - 1.190

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR