Mangansulfat monohydrat 99.0-101.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate

25303.233EA 337 DKK
25303.233 25303.290 25303.460
Mangansulfat monohydrat 99.0-101.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Mangansulfat monohydrat
Formel: MnSO₄·1H₂O
Molekylvægt: 169.02 g/mol
Smeltepunkt: 700
Massefylde: 2.95 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149159
CAS-nummer: 10034-96-5
EINECS: 232-089-9
UN nummer: 3077
ADR: 9 III
Merck Index: 13,05763

Bestil nu

Specifikationer Testresultater

Assay 99.0 - 101.0 %
Insolubility in water Max. 100 ppm
Loss on ignition (500°C) 10.0 - 12.0 %
Substances reducing KMnO4 Passes test
Cl (Chloride) Max. 10 ppm
Ca (Calcium) Max. 50 ppm
Heavy metals (as Pb) Max. 10 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 50 ppm
Na (Sodium) Max. 50 ppm
Ni (Nickel) Max. 0.02 %
Pb (Lead) Max. 10 ppm
Zn (Zinc) Max. 50 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR