Lithiumkarbonat 99,0, AnalaR NORMAPUR® Reag. Ph. Eur. zA

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

25007.230EA 765 DKK
25007.230
Lithiumkarbonat 99,0, AnalaR NORMAPUR® Reag. Ph. Eur. zA
Lithiumkarbonat
Formel: Li₂CO₃
Molekylvægt: 73.89 g/mol
Smeltepunkt: 720
Massefylde: 2.11 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011084
CAS-nummer: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Bestil nu

Specifikationer Testresultater

Assay Min. 99.0 %
Heavy metals (as Pb) Max. 10 ppm
Insolubility in hydrochloric acid Max. 100 ppm
Cl (Chloride) Max. 20 ppm
NH4 (Ammonium) Max. 5 ppm
NO3 (Nitrate) Max. 5 ppm
SO4 (Sulphate) Max. 0.02 %
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 10 ppm
Na (Sodium) Max. 50 ppm
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR